Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105371303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003020.1 | XP_016858509.1 | 4 | 584 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538005.1 | - | 4 | 568 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 2 | 166 | ||
NM_173638.4 | NP_775909.2 | 1 | 67 | |||
GenBank mRNA | AK097180.1 | - | 2 | 85 | ||
BQ890458.1 | - | 3 | 361 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308678.1 | - | 2 | 339 |