Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIAL4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164262.2 | NP_001157734.2 | 1 | 458 |
Gene Symbol: PPIAL4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164261.1 | NP_001157733.1 | 1 | 458 |
Gene Symbol: PPIAL4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144032.2 | NP_001137504.2 | 1 | 458 |