Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098670.1 | NP_001092140.1 | 5 | 542 | ||
NM_001098671.1 | NP_001092141.1 | 5 | 582 | |||
NM_001318398.1 | NP_001305327.1 | 5 | 502 | |||
NM_153819.1 | NP_722541.1 | 5 | 608 | |||
XM_005273707.3 | XP_005273764.3 | 5 | 933 | |||
XM_011544718.1 | XP_011543020.1 | 5 | 933 | |||
XM_011544720.1 | XP_011543022.1 | 5 | 1013 | |||
XM_011544721.1 | XP_011543023.1 | 5 | 545 | |||
XM_011544722.1 | XP_011543024.1 | 5 | 598 | |||
XM_011544723.2 | XP_011543025.1 | 5 | 547 | |||
XM_011544725.2 | XP_011543027.1 | 3 | 297 | |||
XM_017017082.1 | XP_016872571.1 | 4 | 1583 | |||
XM_017017083.1 | XP_016872572.1 | 4 | 1583 | |||
XM_017017084.1 | XP_016872573.1 | 5 | 576 | |||
XM_017017085.1 | XP_016872574.1 | 2 | 366 | |||
GenBank mRNA | AF043722.1 | - | 6 | 714 | ||
AF043723.1 | - | 5 | 608 | |||
AF081194.1 | - | 5 | 515 | |||
AK092882.1 | - | 4 | 1278 | |||
AK122694.1 | - | 5 | 466 | |||
AK124023.1 | - | 4 | 2078 | |||
AK314637.1 | - | 4 | 493 | |||
BC110306.1 | - | 5 | 516 | |||
BC117151.1 | - | 5 | 606 | 40125702 | ||
DA529151.1 | - | 5 | 582 | |||
U78170.1 | - | 5 | 515 | |||
Y12336.1 | - | 5 | 608 |