Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515396.1 | XP_011513698.1 | 1 | 255 | ||
XM_017012180.1 | XP_016867669.1 | 1 | 255 | |||
XM_017012181.1 | XP_016867670.1 | 1 | 255 | |||
GenBank mRNA | AK295044.1 | - | 1 | 255 |