Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278911.1 | NP_001265840.1 | 4 | 626 | ||
NM_005832.4 | NP_005823.1 | 5 | 703 | |||
NM_181361.2 | NP_852006.1 | 4 | 755 | |||
XM_011512325.2 | XP_011510627.1 | 5 | 736 | |||
GenBank mRNA | AA904191.1 | - | 5 | 541 | ||
AB193524.1 | - | 4 | 217 | |||
AF099137.1 | - | 4 | 419 | |||
AF209747.1 | - | 4 | 626 | |||
AK298291.1 | - | 4 | 595 | |||
BC017825.1 | - | 4 | 617 | 4657825 | ||
BG701449.1 | - | 4 | 766 |