Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT17P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001443.1 | 1 | 2647 | |||
GenBank mRNA | AK127974.1 | - | 8 | 2065 | ||
BC040601.1 | - | 7 | 2283 |
Gene Symbol: KRT42P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033415.1 | 11 | 2593 | |||
GenBank mRNA | AK095281.1 | - | 9 | 2624 | ||
AK125965.1 | - | 11 | 2593 |