Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 3 | 609 | ||
GenBank mRNA | AB307689.1 | - | 1 | 117 | ||
AK290588.1 | - | 3 | 605 | |||
AL701206.1 | - | 3 | 551 | |||
BC093727.1 | - | 1 | 141 | 7939572 | ||
BC093729.1 | - | 1 | 141 | 7939574 | ||
HQ692828.1 | - | 1 | 117 | |||
M24857.1 | - | 3 | 531 | |||
M38258.1 | - | 1 | 263 | |||
M57707.1 | - | 2 | 316 |