Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005836.2 | NP_005827.1 | 3 | 281 | ||
GenBank mRNA | BC010280.1 | - | 3 | 249 | 3048297 | |
BC012592.1 | - | 3 | 282 | 4102470 | ||
BC093059.1 | - | 3 | 262 | 30370716 | ||
CR456844.1 | - | 3 | 187 | |||
CR541652.1 | - | 3 | 187 | |||
X95384.1 | - | 3 | 281 |