Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005837.2 | NP_005828.2 | 2 | 389 | ||
GenBank mRNA | BC001430.2 | - | 2 | 299 | 3138336 | |
BI666854.1 | - | 2 | 393 | |||
U94316.1 | - | 2 | 295 |