Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FFAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1113 | ||
XM_011526858.2 | XP_011525160.1 | 2 | 1469 | |||
GenBank mRNA | AK290710.1 | - | 2 | 1113 | ||
BC035657.1 | - | 2 | 1099 | 5588752 | ||
BC113695.1 | - | 1 | 1108 | 8322747 |