Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303481.1 | NP_001290410.1 | 9 | 3206 | ||
NM_003575.3 | NP_003566.1 | 8 | 3463 | |||
XM_006716151.3 | XP_006716214.1 | 8 | 3460 | |||
GenBank mRNA | BC009542.1 | - | 1 | 306 | ||
BC031686.1 | - | 4 | 2218 | |||
BC041704.1 | - | 8 | 3190 | |||
BC055287.1 | - | 8 | 3255 |