Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005843.5 | NP_005834.4 | 12 | 1463 | ||
GenBank mRNA | AF042273.1 | - | 12 | 1222 | ||
AK292265.1 | - | 12 | 1158 | |||
AK292847.1 | - | 12 | 1200 | |||
AK310863.1 | - | 12 | 1200 | |||
AL133600.1 | - | 12 | 1225 | |||
BC028740.2 | - | 12 | 1463 | 5260998 |