Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297647.1 | NP_001284576.1 | 6, 7 | 686 | ||
NM_001297648.1 | NP_001284577.1 | 6, 7 | 681 | |||
NM_006314.2 | NP_006305.2 | 6, 7 | 686 | |||
XM_011540484.2 | XP_011538786.1 | 6, 7 | 666 | |||
XM_011540485.2 | XP_011538787.1 | 3, 4 | 175 | |||
GenBank mRNA | AB209335.1 | - | 6, 7 | 681 | ||
AF100153.1 | - | 6, 7 | 664 | |||
AK001900.1 | - | 6, 7 | 660 | |||
AK222817.1 | - | 6, 7 | 665 | |||
AK222904.1 | - | 6, 7 | 669 | |||
AK292646.1 | - | 6, 7 | 668 | |||
AK296814.1 | - | 6, 7 | 648 | |||
AK302056.1 | - | 6, 7 | 667 | |||
BC011604.2 | - | 6, 7 | 667 | 3139227 | ||
BC012797.2 | - | 6, 7 | 667 | 3505293 | ||
BT006900.1 | - | 6, 7 | 628 | |||
DR159101.1 | - | 6, 7 | 629 |