Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198082.2 | NP_932348.2 | 17 | 2732 | ||
XM_017024463.1 | XP_016879952.1 | 21 | 3460 | |||
XM_017024466.1 | XP_016879955.1 | 20 | 3286 | |||
XM_017024479.1 | XP_016879968.1 | 19 | 3476 | |||
XM_017024483.1 | XP_016879972.1 | 17 | 2941 | |||
GenBank mRNA | BC110997.1 | - | 18 | 2735 | 5725904 |