Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144823.2 | NP_001138295.1 | 11 | 1850 | ||
NM_001320835.1 | NP_001307764.1 | 11 | 1850 | |||
NM_005848.4 | NP_005839.3 | 11 | 1843 | |||
XM_005254120.4 | XP_005254177.1 | 10 | 1629 | |||
XM_005254121.3 | XP_005254178.1 | 11 | 1850 | |||
XM_005254122.4 | XP_005254179.1 | 11 | 1709 | |||
XM_006720371.3 | XP_006720434.1 | 11 | 1585 | |||
XM_011521156.2 | XP_011519458.1 | 11 | 1845 | |||
XM_017021862.1 | XP_016877351.1 | 10 | 1570 | |||
XM_017021863.1 | XP_016877352.1 | 10 | 1628 | |||
XM_017021864.1 | XP_016877353.1 | 11 | 1845 | |||
XM_017021865.1 | XP_016877354.1 | 11 | 1708 | |||
GenBank mRNA | AF534403.1 | - | 11 | 1805 | ||
AK091368.1 | - | 11 | 1805 | |||
AK290292.1 | - | 11 | 1658 | |||
AK299570.1 | - | 11 | 1764 | |||
AK307411.1 | - | 11 | 1850 | |||
AL832602.2 | - | 9 | 1369 | |||
AL833317.1 | - | 11 | 1681 | |||
AL833384.1 | - | 8 | 1114 | |||
BC028057.1 | - | 11 | 1695 | |||
BC041706.1 | - | 11 | 1674 | 5530033 |