Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271849.1 | NP_001258778.1 | 2 | 387 | ||
NM_005851.4 | NP_005842.1 | 2 | 787 | |||
NR_073484.1 | 1 | 557 | ||||
GenBank mRNA | AF089814.1 | - | 2 | 244 | ||
AK001498.1 | - | 2 | 221 | |||
AK310197.1 | - | 2 | 358 | |||
BC002850.2 | - | 2 | 690 | 3659410 | ||
BC016704.1 | - | 2 | 226 | 4075420 | ||
BI763791.1 | - | 1 | 408 | |||
BM559572.1 | - | 503 | ||||
BT006909.1 | - | 2 | 141 | |||
CB988039.1 | - | 1 | 584 | |||
CR457050.1 | - | 2 | 141 | |||
CR542042.1 | - | 2 | 141 |