Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271849.1 | NP_001258778.1 | 4 | 1036 | ||
NM_005851.4 | NP_005842.1 | 4 | 1436 | |||
NR_073484.1 | 3 | 1206 | ||||
GenBank mRNA | AF089814.1 | - | 4 | 893 | ||
AK001498.1 | - | 4 | 870 | |||
BC002850.2 | - | 4 | 1339 | 3659410 | ||
BC016704.1 | - | 4 | 875 | 4075420 |