Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 1054 | |||
GenBank mRNA | AY894575.1 | - | 7 | 936 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 3 | 8964 | ||
XM_017030107.1 | XP_016885596.1 | 3 | 8964 |