Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 5 | 1373 | ||
XM_011515604.2 | XP_011513906.1 | 6 | 1559 | |||
XM_011515608.2 | XP_011513910.1 | 6 | 1463 | |||
XM_011515609.2 | XP_011513911.1 | 4 | 1354 | |||
XM_011515610.2 | XP_011513912.1 | 7 | 1640 | |||
XM_011515611.2 | XP_011513913.1 | 5 | 1507 | |||
XM_017012777.1 | XP_016868266.1 | 4 | 1208 | |||
XM_017012778.1 | XP_016868267.1 | 5 | 1411 | |||
GenBank mRNA | AF277624.1 | - | 5 | 1373 |