Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 36 | 4289 | ||
XM_005261634.1 | XP_005261691.1 | 35 | 4196 | |||
XM_005261635.1 | XP_005261692.1 | 36 | 4289 | |||
XM_017028829.1 | XP_016884318.1 | 35 | 4223 | |||
XM_017028830.1 | XP_016884319.1 | 22 | 2445 | |||
GenBank mRNA | AB210002.1 | - | 36 | 4259 | ||
AF012872.1 | - | 35 | 4028 | |||
AK302424.1 | - | 36 | 4217 | |||
AK314781.1 | - | 8 | 943 | |||
BC018120.1 | - | 6 | 606 | |||
BC053654.1 | - | 5 | 582 | |||
L36151.1 | - | 4 | 575 |
Gene Symbol: PI4KAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003563.1 | 4 | 461 | |||
GenBank mRNA | AK096724.1 | - | 6 | 687 | ||
AK292695.1 | - | 5 | 552 | |||
BC072398.1 | - | 6 | 737 | |||
BC131538.1 | - | 4 | 461 |
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003700.1 | 6 | 728 | |||
GenBank mRNA | AK126860.1 | - | 4 | 2692 | ||
AK298744.1 | - | 6 | 749 | |||
AK299734.1 | - | 6 | 593 | |||
BC020225.1 | - | 6 | 726 | |||
BC139841.1 | - | 4 | 449 | |||
BX452846.2 | - | 6 | 788 | |||
CR456378.1 | - | 4 | 467 | |||
CX751888.1 | - | 1 | 33 |