Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005856.2 | NP_005847.1 | 3 | 431 | ||
XM_006715631.3 | XP_006715694.1 | 5 | 754 | |||
XM_017011666.1 | XP_016867155.1 | 3 | 795 | |||
GenBank mRNA | AJ001016.1 | - | 3 | 399 | ||
AK128765.1 | - | 1 | 1761 | |||
AK222469.1 | - | 3 | 395 | |||
AK311871.1 | - | 3 | 370 | |||
AY265459.1 | - | 3 | 370 | |||
BC022304.1 | - | 3 | 398 | 4717934 | ||
BC053852.1 | - | 3 | 397 | 6197363 | ||
BP347072.1 | - | 3 | 431 |