Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161797.1 | NP_001155269.1 | 9, 10 | 1845 | ||
NM_175306.4 | NP_780515.2 | 10, 11 | 1926 | |||
GenBank mRNA | AK049209.1 | - | 10, 11 | 1889 | ||
AK220496.1 | - | 9, 10 | 1824 | |||
BC075672.1 | - | 10, 11 | 1888 | 30549496 | ||
BC096033.1 | - | 10, 11 | 1845 | 6854767 |