Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190733.1 | NP_001177662.1 | 7 | 586 | ||
NM_001190734.1 | NP_001177663.1 | 8 | 521 | |||
NM_178896.5 | NP_849227.1 | 7 | 586 | |||
GenBank mRNA | AK033559.1 | - | 7 | 531 | ||
AK076991.1 | - | 8 | 522 | |||
BC060213.1 | - | 7 | 528 | |||
BC108346.1 | - | 7 | 488 | 6310392 | ||
BC137627.1 | - | 7 | 530 | 8860646 | ||
BC137628.1 | - | 7 | 530 | 8860647 | ||
BC144794.1 | - | 7 | 530 | 9053342 | ||
CO433728.1 | - | 3 | 195 |