Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133937.1 | NP_598698.1 | 1 | 71 | ||
GenBank mRNA | AK003342.1 | - | 1 | 86 | ||
AK020128.1 | - | 1 | 86 | |||
AK020277.1 | - | 1 | 87 | |||
AK150689.1 | - | 1 | 86 | |||
AK163236.1 | - | 1 | 86 | |||
AK166585.1 | - | 1 | 74 | |||
BC010582.1 | - | 1 | 71 | 3154777 |