Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hinfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172162.3 | NP_751894.2 | 3, 4 | 474 | ||
GenBank mRNA | AK049348.1 | - | 3, 4 | 438 | ||
AK049633.1 | - | 3, 4 | 635 | |||
BC029091.1 | - | 2, 3 | 457 | 2651006 | ||
CN708951.1 | - | 2, 3 | 320 | |||
CT010239.1 | - | 2, 3 | 409 |