Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028821.3 | NP_083097.2 | 6 | 445 | ||
GenBank mRNA | AK005826.1 | - | 6 | 531 | ||
BC070454.1 | - | 402 | ||||
BC125392.1 | - | 5 | 427 | 40129907 | ||
BC125394.1 | - | 5 | 423 | 40129909 |