Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175341.4 | NP_780550.1 | 3 | 993 | ||
NM_207515.2 | NP_997398.1 | 3 | 993 | |||
GenBank mRNA | AK028797.1 | - | 3 | 958 | ||
AK085709.1 | - | 3 | 956 | |||
AK151913.1 | - | 3 | 957 | |||
AK151926.1 | - | 3 | 957 | |||
AK151957.1 | - | 3 | 957 | |||
AK153261.1 | - | 3 | 847 | |||
AK159213.1 | - | 3 | 951 | |||
AK164372.1 | - | 3 | 956 | |||
AK167479.1 | - | 3 | 823 | |||
AK167694.1 | - | 4 | 759 | |||
AK177332.1 | - | 2 | 236 | |||
AK192385.1 | - | 1 | 134 | |||
AK220470.1 | - | 967 | ||||
BC020406.1 | - | 2 | 391 | |||
BC023895.1 | - | 3 | 945 | |||
BC026711.1 | - | 2 | 358 | |||
BC075665.1 | - | 2 | 596 | 30605146 | ||
BC079898.1 | - | 3 | 942 | |||
GQ906534.1 | - | 2 | 311 |