Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153083.5 | NP_694723.1 | 1 | 1106 | ||
GenBank mRNA | AF432864.2 | - | 1 | 937 | ||
AK085173.1 | - | 1 | 801 | |||
AK132479.1 | - | 1 | 798 | |||
BC025562.1 | - | 1 | 1090 | 5322971 |