Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029128.2 | NP_083404.2 | 3 | 373 | ||
GenBank mRNA | AK014045.1 | - | 3 | 518 | ||
AK015536.1 | - | 411 | ||||
BC017628.1 | - | 3 | 345 | |||
BC083089.1 | - | 3 | 353 | 30105859 | ||
FM985972.1 | - | 2 | 253 |