Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146079.1 | NP_666191.1 | 4 | 1037 | ||
GenBank mRNA | AK033052.1 | - | 4 | 1002 | ||
BC018258.1 | - | 4 | 965 | 4511650 | ||
BC018480.1 | - | 4 | 1006 | 4500919 | ||
BC029017.1 | - | 4 | 982 | 5359531 | ||
BC029661.1 | - | 4 | 992 | 5366068 | ||
BC031367.1 | - | 4 | 1000 | 5358667 | ||
BC031916.1 | - | 4 | 1037 | 5369642 | ||
BC033348.1 | - | 4 | 987 | |||
BC033358.1 | - | 4 | 1036 | 5368968 | ||
BC033364.1 | - | 4 | 987 | |||
BC033380.1 | - | 4 | 992 | 5366953 | ||
BC033383.1 | - | 4 | 993 | 5368064 | ||
BC033396.1 | - | 4 | 984 | |||
BC033423.1 | - | 4 | 989 | |||
BC034331.1 | - | 4 | 993 | 5357797 | ||
BC034595.1 | - | 4 | 992 | 5362191 |