Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030693.2 | NP_109618.1 | 3 | 1499 | ||
GenBank mRNA | AB012276.1 | - | 1 | 609 | ||
AF310624.1 | - | 3 | 1520 | |||
AF312938.1 | - | 3 | 1475 | |||
AF375475.1 | - | 3 | 1504 | |||
AF375476.1 | - | 3 | 1499 | |||
AK131939.1 | - | 3 | 1521 | |||
AK146479.1 | - | 3 | 1521 | |||
AK159624.1 | - | 3 | 1523 | |||
AK160085.1 | - | 1 | 924 | |||
BC010195.1 | - | 2 | 1332 | 3157499 | ||
BC092068.1 | - | 3 | 1504 | 4210931 |