Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 2 | 209 | ||
GenBank mRNA | AK161028.1 | - | 2 | 293 | ||
AY237726.1 | - | 2 | 238 | |||
BC058610.1 | - | 2 | 221 | 30056142 | ||
BC145279.1 | - | 2 | 196 | 9053833 | ||
BC150749.1 | - | 2 | 196 | 9087660 | ||
BC171954.1 | - | 2 | 196 |