Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025315.3 | NP_079591.1 | 3 | 222 | ||
GenBank mRNA | AK009742.1 | - | 3 | 223 | ||
AK012217.1 | - | 3 | 223 | |||
AK014428.1 | - | 3 | 216 | |||
AK019417.1 | - | 3 | 212 | |||
BC012286.1 | - | 3 | 213 | 3493056 | ||
BC096641.1 | - | 2 | 1172 |