Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025315.3 | NP_079591.1 | 1, 2 | 75 | ||
GenBank mRNA | AK009742.1 | - | 1, 2 | 76 | ||
AK012217.1 | - | 1, 2 | 76 | |||
AK014428.1 | - | 1, 2 | 69 | |||
AK019417.1 | - | 1, 2 | 65 | |||
BC012286.1 | - | 1, 2 | 66 | 3493056 |