Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133998.3 | NP_598759.1 | 2 | 260 | ||
GenBank mRNA | AF391115.1 | - | 2 | 233 | ||
AK076516.1 | - | 2 | 231 | |||
AK088306.1 | - | 2 | 228 | |||
AK160157.1 | - | 2 | 131 | |||
BB840302.1 | - | 2 | 261 | |||
BC034216.1 | - | 2 | 206 | 5008732 |