Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175414.4 | NP_780623.1 | 9 | 1214 | ||
GenBank mRNA | AK079139.1 | - | 9 | 1187 | ||
AK158729.1 | - | 8 | 1099 | |||
AK163395.1 | - | 9 | 1270 | |||
BC026201.1 | - | 1 | 325 | |||
BC052503.1 | - | 9 | 1196 | 5146854 |