Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177231.2 | NP_796205.1 | 8 | 867 | ||
NM_178220.3 | NP_835738.1 | 8 | 867 | |||
GenBank mRNA | AK052588.1 | - | 8 | 644 | ||
AK090090.1 | - | 8 | 694 | |||
AK144054.1 | - | 8 | 666 | |||
AK147508.1 | - | 8 | 868 | |||
AK147566.1 | - | 8 | 889 | |||
AK165514.1 | - | 8 | 666 | |||
BC108969.1 | - | 8 | 634 | 40047224 | ||
BC108970.1 | - | 8 | 634 | 40047225 |