Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 4, 5 | 536 | ||
GenBank mRNA | AK007546.1 | - | 1, 2 | 85 | ||
AK012615.1 | - | 4, 5 | 477 | |||
AK012877.1 | - | 4, 5 | 471 | |||
AK033451.1 | - | 4, 5 | 426 | |||
AK154013.1 | - | 4, 5 | 518 | |||
BC048844.1 | - | 4, 5 | 486 | |||
BC055451.1 | - | 4, 5 | 482 | 4164972 |