Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081274.1 | NP_001074743.1 | 3 | 354 | ||
GenBank mRNA | AK002894.1 | - | 3 | 316 | ||
AK077110.1 | - | 3 | 316 | |||
AK145602.1 | - | 3 | 318 | |||
AK150210.1 | - | 3 | 317 | |||
AK150490.1 | - | 3 | 313 | |||
AK151380.1 | - | 3 | 313 | |||
AK151993.1 | - | 3 | 313 | |||
AK152758.1 | - | 3 | 313 | |||
AK152894.1 | - | 3 | 313 | |||
AK153409.1 | - | 3 | 310 | |||
AK155027.1 | - | 3 | 313 | |||
AK161869.1 | - | 3 | 312 | |||
AK166733.1 | - | 3 | 308 | |||
AK166947.1 | - | 3 | 290 | |||
AK167215.1 | - | 3 | 308 | |||
AK168251.1 | - | 3 | 313 | |||
BC008646.1 | - | 3 | 285 | 3155147 | ||
BC011329.1 | - | 3 | 303 | 3984654 | ||
BC014793.1 | - | 3 | 322 | 3155100 |