Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 1236 | ||
NM_173374.4 | NP_775550.2 | 4 | 1040 | |||
GenBank mRNA | AK053674.1 | - | 4 | 731 | ||
AK078715.1 | - | 3 | 893 | |||
AK150535.1 | - | 4 | 743 | |||
AK169203.1 | - | 4 | 697 | |||
AK169623.1 | - | 4 | 698 | |||
BC046773.1 | - | 4 | 672 | 5708288 | ||
BC058627.1 | - | 4 | 706 | 6401674 | ||
X66091.1 | - | 3 | 849 |