Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053259.2 | NP_444489.2 | 6 | 939 | ||
GenBank mRNA | AF184895.1 | - | 5 | 778 | ||
AK144498.1 | - | 6 | 940 | |||
BC119386.1 | - | 5 | 806 | 8734135 | ||
BC119388.1 | - | 5 | 806 | 8734137 |