Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301230.1 | NP_001288159.1 | 5 | 674 | ||
NM_001301232.1 | NP_001288161.1 | 4 | 567 | |||
NM_023635.6 | NP_076124.1 | 4 | 525 | |||
GenBank mRNA | AB046693.1 | - | 3 | 323 | ||
AB120511.1 | - | 3 | 301 | |||
AB232622.1 | - | 3 | 301 | |||
AF304376.1 | - | 4 | 480 | |||
AK010373.1 | - | 4 | 443 | |||
AK030529.1 | - | 4 | 472 | |||
BC008173.1 | - | 5 | 631 | 3710213 | ||
BC009656.1 | - | 4 | 478 | 3590089 | ||
CV784419.1 | - | 4 | 436 |