Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002718.1 | - | 2 | 172 | ||
AK008346.1 | - | 2 | 170 | |||
AK011886.1 | - | 2 | 169 | |||
AK152389.1 | - | 3 | 1187 | |||
AK161394.1 | - | 1 | 261 | |||
AV462016.1 | - | 2 | 233 | |||
BC012680.1 | - | 2 | 231 | 4217188 | ||
BU055998.1 | - | 2 | 158 | |||
CB848021.2 | - | 2 | 195 |