Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039467.1 | NP_001034556.1 | 6 | 660 | ||
NM_005873.2 | NP_005864.1 | 6 | 760 | |||
XM_005260182.3 | XP_005260239.1 | 5 | 1008 | |||
XM_005260183.2 | XP_005260240.1 | 5 | 595 | |||
XM_011528484.1 | XP_011526786.1 | 5 | 2190 | |||
XM_011528485.2 | XP_011526787.1 | 6 | 1239 | |||
XM_011528486.2 | XP_011526788.1 | 6 | 692 | |||
GenBank mRNA | AF493939.1 | - | 5 | 493 | ||
AK290081.1 | - | 6 | 777 | |||
AK298240.1 | - | 675 | ||||
AY585188.1 | - | 5 | 493 | |||
BC001318.1 | - | 5 | 575 | 3454625 | ||
BC054337.1 | - | 6 | 703 | 6067115 | ||
BC063010.1 | - | 6 | 748 | 5215492 | ||
BT009804.1 | - | 5 | 493 | |||
CR457430.1 | - | 5 | 493 | |||
X91809.1 | - | 6 | 781 |