Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301206.1 | NP_001288135.1 | 4 | 1845 | ||
NM_001717.3 | NP_001708.3 | 4 | 1709 | |||
XM_011521893.1 | XP_011520195.1 | 4 | 1611 | |||
XM_011521894.1 | XP_011520196.1 | 3 | 1527 | |||
GenBank mRNA | AK302992.1 | - | 5 | 1725 | ||
DB051299.1 | - | 1 | 222 | |||
L03427.1 | - | 1705 |