Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013482.2 | NP_038510.2 | 6 | 567 | ||
GenBank mRNA | L08967.1 | - | 6 | 543 | ||
L10627.1 | - | 6 | 572 | |||
L29788.1 | - | 6 | 539 |