Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284508.1 | NP_001271437.1 | 4 | 697 | ||
NM_031161.4 | NP_112438.1 | 3 | 534 | |||
GenBank mRNA | AK002677.1 | - | 3 | 502 | ||
BC028487.1 | - | 4 | 642 | 1400830 | ||
M11739.1 | - | 2 | 273 |