Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007631.2 | NP_031657.1 | 1 | 285 | ||
GenBank mRNA | AK017834.1 | - | 1 | 249 | ||
AK053348.1 | - | 1 | 193 | |||
AK149905.1 | - | 1 | 192 | |||
AK164997.1 | - | 1 | 230 | |||
BC044841.1 | - | 1 | 188 | 3155470 | ||
BC060172.1 | - | 1 | 180 | |||
BY089102.1 | - | 1 | 285 | |||
EU600170.1 | - | 1 | 285 | |||
M64403.1 | - | 1 | 240 | |||
S78355.1 | - | 1 | 190 |