Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005875.2 | NP_005866.1 | 4 | 570 | ||
GenBank mRNA | AF064607.1 | - | 4 | 380 | ||
AF077052.1 | - | 4 | 551 | |||
AF263452.1 | - | 4 | 543 | |||
AK223448.1 | - | 4 | 565 | |||
AK311922.1 | - | 4 | 475 | |||
BC006996.1 | - | 4 | 554 | 4044439 | ||
CA335728.1 | - | 4 | 583 | |||
CR542223.1 | - | 4 | 310 |