Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287738.1 | NP_001274667.1 | 1 | 1286 | ||
NM_001287739.1 | NP_001274668.1 | 1 | 1286 | |||
NM_009883.4 | NP_034013.1 | 1 | 1286 | |||
GenBank mRNA | AK143374.1 | - | 1 | 1235 |